Learn More
Invitrogen™ PRDM9 Chimeric Recombinant Rabbit Monoclonal Antibody (RAB-C370)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA548336
Description
Specificity: This antibody recognizes PRDM9 (Histone-lysine N-methyltransferase PRDM9). It binds to a folded domain, amino acids 195-385. PRDM9 is a histone methyltransferase that plays a key role in meiotic prophase by determining hotspot localization thereby promoting meiotic recombination.
The PR domain is a protein-protein interaction module of about 100 amino acids. PR domain-containing proteins, such as PRDM9, are often involved in transcriptional regulation.
Specifications
PRDM9 Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
Q9NQV7 | |
PRDM9 | |
This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRDM9 protein under non-denaturing conditions. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG κ |
ChIP Assay, ELISA, Flow Cytometry, Immunoprecipitation, Immunocytochemistry | |
RAB-C370 | |
Unconjugated | |
PRDM9 | |
Histone-lysine N-methyltransferase PRDM9; MEISETZ; minisatellite binding protein 3 (115kD); MSBP3; PFM6; PR domain 9; PR domain containing 7; PR domain containing 9; PR domain zinc finger protein 9; PR domain-containing protein 9; Prdm7; Prdm9; Protein-lysine N-methyltransferase PRDM9; ZNF899 | |
Rabbit | |
Protein A | |
RUO | |
56979 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.