Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584424
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584424 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584424 Supplier Invitrogen™ Supplier No. PA584424
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QELGIKWGSK WKKELMAGRE PKPEIHPCPS CCLAFSSQKF LSQHVERNHS SQNFPGPSAR KLLQPENPCP G Highest antigen sequence indentity to the following orthologs: Mouse - 63%, Rat - 66%.

The PR domain is a protein-protein interaction module of about 100 amino acids. PR domain-containing proteins, such as PRDM9, are often involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRDM9
Gene Accession No. Q9NQV7
Gene Alias Histone-lysine N-methyltransferase PRDM9; MEISETZ; minisatellite binding protein 3 (115kD); MSBP3; PFM6; PR domain 9; PR domain containing 7; PR domain containing 9; PR domain zinc finger protein 9; PR domain-containing protein 9; Prdm7; Prdm9; Protein-lysine N-methyltransferase PRDM9; ZNF899
Gene Symbols PRDM9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRDM9. Recombinant protein control fragment (Product #RP-101424).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56979
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.