Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDM9, Rabbit anti-Mouse, Human, Polyclonal, Millipore Sigma™

Catalog No. ABE194725UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE194725UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE194725UG Supplier MilliporeSigma Supplier No. ABE194725UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-PRDM9, Cat. No. ABE1947, is a highly specific rabbit polyclonal antibody that targets Histone-lysine N-methyltransferase PRDM9 in human and murine cells and has been tested for use in Western Blotting.

Histone-lysine N-methyltransferase PRDM9 (UniProt: Q9NQV7; EC: 2.1.1.43, PR domain zinc finger protein 9, PR domain-containing protein 9, PRDM9) is encoded by PRDM9 (also known as PFM6) gene (Gene ID: 56979) in human. PRDM9 is a zinc-finger protein that catalyzes histone H3 Lysine 4 trimethylation (H3K4me3), but lacks capacity to mono- and dimethylate Lysine 4 of histone H3. H3 Lysine -4 methylation plays a vital role in the transcriptional activation of genes during early meiotic prophase. PRDM9 contains multiple domains, including a Kruppel-associated box (KRAB) domain (aa 23-86), an SSX repression domain (SSXRD), a PRD1-BF1 and RIZ homologous region, a subclass of SET (PR/SET) domain (aa 244-358), and a tandem array of C2H2 zinc fingers. The zinc finger array recognizes a short sequence motif, leading to local H3K4me3, and meiotic recombination hotspot activity. PRDM9 is reported to be highly polymorphic, both within and between mammalian species. It is shown that most of the naturally occurring sequence polymorphisms in PRDM9 change the identity of the amino acids contacting DNA and/or the number and arrangement of individual fingers in the DNA-binding zinc-finger domains. This allows PRDM9 variants to target a large number of DNA sequences, thereby expanding the distribution of recombination sites. (Ref.: Baker, CL et al. (2015). PLoS Genet 11(9): e1005512).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blotting Analysis: A representaive lot detected PRDM9 in Wild type, but not in KO mouse testis tissue lysate ( Courtesy  of Guccione Lab @ A*STAR( Institute of Molecular Cell Biology, IMCB) ).
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Histone-lysine N-methyltransferase PRDM9;EC: 2.1.1.43;PR domain zinc finger protein 9;PR domain-containing protein 9
Gene Symbols PRDM9;PFM6
Host Species Rabbit
Immunogen GST-tagged recombinant fragment corresponding to 191 amino acids from the N-terminal half of human Histone-lysine N-methyltransferase PRDM9.
Molecular Weight of Antigen ∽90 kda observed; 103.37 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Histone-lysine N-methyltransferase PRDM9 in human and mouse cells. It targets an epitope within 191 amino acids from the N-terminal half.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.