Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDM9 Recombinant Rabbit Monoclonal Antibody (1H9Z1)
GREENER_CHOICE

Catalog No. MA568593
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA568593 20 μL
FEMA551196 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA568593 Supplier Invitrogen™ Supplier No. MA568593
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: DSEDSDEEWT PKQQVSPPWV PFRVKHSKQQ KESSRMPFSG ESNVKEGSGI ENLLNTSGSE HVQKPVSSLE EGNTSGQHSG KKLKLRKKNV EVKMYRLRER KGLAYEEVSE PQDDDYLYCE KCQNFFIDSC PNHGPPLFVK DSMVDRGHPN HSVLSLPPGL RISPSGIPEA GLGVWNEASD LPVGLHFGPY EGQITEDEEA ANSGYSWLIT KGRNCYEYVD GQDESQANWM RYVNCARDDE EQNLVAFQYH RKIFYRTCRV IRPGCELLVW YGDEYGQELG IKWG.

The PR domain is a protein-protein interaction module of about 100 amino acids. PR domain-containing proteins, such as PRDM9, are often involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDM9
Applications ELISA, Immunohistochemistry (Paraffin), CUT&RUN
Classification Recombinant Monoclonal
Clone 1H9Z1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene PRDM9
Gene Accession No. Q9NQV7
Gene Alias Histone-lysine N-methyltransferase PRDM9; MEISETZ; minisatellite binding protein 3 (115kD); MSBP3; PFM6; PR domain 9; PR domain containing 7; PR domain containing 9; PR domain zinc finger protein 9; PR domain-containing protein 9; Prdm7; Prdm9; Protein-lysine N-methyltransferase PRDM9; ZNF899
Gene Symbols PRDM9
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56979
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.