Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prdx-6 Mouse anti-Human, Clone: TAG-2A12, MilliporeSigma™

Catalog No. MABN1797MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN1797MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1797MI Supplier MilliporeSigma Supplier No. MABN1797
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Prdx-6 Antibody, clone TAG-2A12 is a highly specific mouse monoclonal antibody that targets Prdx-6 and has been tested in Flow Cytometry, Immunofluorescence, Immunocytochemistry and Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prdx-6
Applications Flow Cytometry, Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone TAG-2A12
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols PRDX6,AOP2,KIAA0106
Host Species Mouse
Immunogen Corneal endothelial cells (hCENC) isolated from corneas of human cadavers and immunized with monophosphoryl-lipid A trehalose dicorynomycolate (MPLCTDM) adjuvant.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_004896
Test Specificity Clone TAG-2A12 detects Peroxiredoxin-6 in human cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.