Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527487
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527487 Supplier Invitrogen™ Supplier No. PA527487
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, NIH-3T3. Predicted reactivity: Mouse (95%), Rat (98%), Zebrafish (80%), Dog (97%), Chicken (86%), Rhesus Monkey (98%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. Three transcript variants encoding the same protein have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDX1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene PRDX1
Gene Accession No. P35700, Q06830, Q63716
Gene Alias ACR1; Alu co-repressor 1; AOEB166; B166; HBP23; heme-binding 23 kDa protein; macrophage 23 Kd stress protein; macrophage 23 kDa stress protein; macrophage 23kDa stress protein; macrophase stress protein 22kDa; macrophase stress protein 23 kd; Msp23; Natural killer cell-enhancing factor A; natural killer-enhancing factor A; NkefA; NKEF-A; OSF3; OSF-3; osteoblast specific factor 3; osteoblast-specific factor 3; PAG; Paga; PAGB; peroxiredoxin 1; peroxiredoxin-1; PRDX1; PrdxI; proliferation-associated gene A; proliferation-associated gene protein; Prx; Prx 1; PRX1; PRXI; TDPX2; TDX2; thioredoxin dependent peroxide reductase 2; thioredoxin peroxidase 2; Thioredoxin-dependent peroxide reductase 2; TPxA; Trx dependent peroxide reductase 2
Gene Symbols PRDX1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PRX I. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117254, 18477, 5052
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.