Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRDX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118189
This item is not returnable.
View return policy
Description
PRDX1 Polyclonal Antibody for Western Blot, IP, ELISA
This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. Three transcript variants encoding the same protein have been identified for this gene.
Specifications
| PRDX1 | |
| Polyclonal | |
| Unconjugated | |
| PRDX1 | |
| ACR1; Alu co-repressor 1; AOEB166; B166; HBP23; heme-binding 23 kDa protein; macrophage 23 Kd stress protein; macrophage 23 kDa stress protein; macrophage 23kDa stress protein; macrophase stress protein 22kDa; macrophase stress protein 23 kd; Msp23; Natural killer cell-enhancing factor A; natural killer-enhancing factor A; NkefA; NKEF-A; OSF3; OSF-3; osteoblast specific factor 3; osteoblast-specific factor 3; PAG; Paga; PAGB; peroxiredoxin 1; peroxiredoxin-1; PRDX1; PrdxI; proliferation-associated gene A; proliferation-associated gene protein; Prx; Prx 1; PRX1; PRXI; TDPX2; TDX2; thioredoxin dependent peroxide reductase 2; thioredoxin peroxidase 2; Thioredoxin-dependent peroxide reductase 2; TPxA; Trx dependent peroxide reductase 2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 5052 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q06830 | |
| PRDX1 | |
| Recombinant Human Peroxiredoxin 1/PRDX1 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction