Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRDX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579862
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Rat Kidney Tissue, HELA Cell, JURKAT Cell, 293T Cell, A549 Cell.
Peroxiredoxin (Prx) is a growing peroxidase family, whose mammalian members have been known to connect with cell proliferation, differentiation, and apoptosis. Many isoforms (about 50 proteins), collected in accordance to the amino acid sequence homology, particularly amino-terminal region containing active site cysteine residue, and the thiol-specific antioxidant activity, distribute throughout all the kingdoms. Among them, mammalian Prx consists of 6 different members grouped into typical 2-Cys, atypical 2-Cys Prx, and 1-Cys Prx. Except Prx VI belonging to 1-Cys Prx subgroup, the other five 2-Cys Prx isotypes have the thioredoxin-dependent peroxidase (TPx) activity utilizing thioredoxin, thioredoxin reductase, and NADPH as a reducing system. Mammalian Prxs are 20 - 30 kilodalton in molecular size and vary in subcellular localization: Prx I, II, and VI in cytosol, Prx III in mitochondria, Prx IV in ER and secretion, Prx V showing complicated distribution including peroxisome, mitochondria and cytosol.
Specifications
PRDX2 | |
Polyclonal | |
Unconjugated | |
PRDX2 | |
AL022839; Band-8; Calpromotin; epididymis secretory sperm binding protein Li 2a; HEL-S-2a; HGNC:9353; MGC4104; Natural killer cell-enhancing factor B; NkefB; NKEF-B; peroxiredoxin 2; peroxiredoxin-2; Prdx2; protector protein; PRP; Prx II-1; PRX2; PRXII; PTX1; Tdpx1; TDX1; thiol specific antioxidant protein; thiol-specific antioxidant 1; thiol-specific antioxidant protein; thioredoxin dependent peroxide reductase 1; thioredoxin peroxidase 1; thioredoxin reductase; thioredoxin-dependent peroxide reductase 1; Torin; TPx; TPX1; TPx-B; TR; Trx dependent peroxide reductase 1; TSA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21672, 29338, 7001 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
P32119, P35704, Q61171 | |
PRDX2 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human Peroxiredoxin 2 (181-198aa DTIKPNVDDSKEYFSKHN). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction