Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRDX2 Recombinant Rabbit Monoclonal Antibody (SAIC-34D-11)
GREENER_CHOICE

Catalog No. PIMA547916
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547916 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547916 Supplier Invitrogen™ Supplier No. MA547916
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: Recognises human PRDX2.

Peroxiredoxin (Prx) is a growing peroxidase family, whose mammalian members have been known to connect with cell proliferation, differentiation, and apoptosis. Many isoforms (about 50 proteins), collected in accordance to the amino acid sequence homology, particularly amino-terminal region containing active site cysteine residue, and the thiol-specific antioxidant activity, distribute throughout all the kingdoms. Among them, mammalian Prx consists of 6 different members grouped into typical 2-Cys, atypical 2-Cys Prx, and 1-Cys Prx. Except Prx VI belonging to 1-Cys Prx subgroup, the other five 2-Cys Prx isotypes have the thioredoxin-dependent peroxidase (TPx) activity utilizing thioredoxin, thioredoxin reductase, and NADPH as a reducing system. Mammalian Prxs are 20 - 30 kilodalton in molecular size and vary in subcellular localization: Prx I, II, and VI in cytosol, Prx III in mitochondria, Prx IV in ER and secretion, Prx V showing complicated distribution including peroxisome, mitochondria and cytosol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDX2
Applications Flow Cytometry, Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone SAIC-34D-11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene PRDX2
Gene Accession No. P32119
Gene Alias AL022839; Band-8; Calpromotin; epididymis secretory sperm binding protein Li 2a; HEL-S-2a; HGNC:9353; MGC4104; Natural killer cell-enhancing factor B; NkefB; NKEF-B; peroxiredoxin 2; peroxiredoxin-2; Prdx2; protector protein; PRP; Prx II-1; PRX2; PRXII; PTX1; Tdpx1; TDX1; thiol specific antioxidant protein; thiol-specific antioxidant 1; thiol-specific antioxidant protein; thioredoxin dependent peroxide reductase 1; thioredoxin peroxidase 1; thioredoxin reductase; thioredoxin-dependent peroxide reductase 1; Torin; TPx; TPX1; TPx-B; TR; Trx dependent peroxide reductase 1; TSA
Gene Symbols PRDX2
Host Species Rabbit
Immunogen Peptide GLFIIDGK, derived from the cellular redox regulator Peroxiredoxin-2, conjugated to KLH.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7001
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.