Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRDX5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580933
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Peroxiredoxin 5/PRDX5.
Peroxiredoxin (Prx) is a growing peroxidase family, whose mammalian members have been known to connect with cell proliferation, differentiation, and apoptosis. Many isoforms (about 50 proteins), collected in accordance to the amino acid sequence homology, containing active site cysteine residue, and the thiol-specific antioxidant activity, distribute throughout all the kingdoms. Among them, mammalian Prx consists of 6 different members grouped into typical 2-Cys, atypical 2-Cys Prx, and 1-Cys Prx. Except Prx VI belonging to 1-Cys Prx subgroup, the other five 2-Cys Prx isotypes have the thioredoxin-dependent peroxidase (TPx) activity utilizing thioredoxin, thioredoxin reductase, and NADPH as a reducing system. Mammalian Prxs are 20-30 kilodalton in molecular size and vary in subcellular localization: Prx I, II, and VI in cytosol, Prx III in mitochondria, Prx IV in ER and secretion, Prx V showing complicated distribution including peroxisome, mitochondria and cytosol.
Specifications
| PRDX5 | |
| Polyclonal | |
| Unconjugated | |
| PRDX5 | |
| ACR1; Alu corepressor 1; Alu co-repressor 1; Antioxidant enzyme B166; AOEB166; AOPP; B166; epididymis secretory protein Li 55; HEL-S-55; Liver tissue 2D-page spot 2D-0014IV; liver tissue 2D-page spot 71B; peroxiredoxin 5; peroxiredoxin 6; peroxiredoxin V; Peroxiredoxin-5, mitochondrial; peroxisomal antioxidant enzyme; peroxisomal membrane protein 20; PLP; Pmp20; PRDX5; Prdx6; Prx5; PrxV; prx-V; SBBI10; Thioredoxin peroxidase PMP20; thioredoxin reductase; TPx type VI | |
| Rabbit | |
| Protein A | |
| RUO | |
| 25824 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| P30044 | |
| PRDX5 | |
| Recombinant Human Peroxiredoxin 5/PRDX5 protein (Met53-Leu214). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction