Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pre-BCR Mouse anti-Human, PE, Clone: HSL2, BD Phrmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569719
Description
Pre-BCR Monoclonal antibody specifically detects Pre-BCR in Human samples. It is validated for Flow cytometry|Intracellular staining (flow cytometry)
Specifications
Pre-BCR | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
613 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG1 κ |
Flow Cytometry, Control | |
HSL2 | |
PE | |
Pre-B Cell Receptor | |
Human Pre-BCR Complexes Purified from Ltk-Pre-BCR Transfectants | |
0.1 mg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction