Learn More
Edvotek™ Pre-Fixed Microscope Slide: Chromosome Spread
SDPSCI_ED
Catalog No. s29006 Shop All EDVOTEK Products
Change view
Click to view available options
Mounting Type:
Whole Mount
Catalog No. Mounting Type
S29006 Whole Mount
1 options
Catalog No. S29006
Supplier: Edvotek™ 987
Only null left
Add to Cart
Add to Cart
Requires
A microscope with 400x magnification
Students will develop an understanding of karyotyping and the association of chromosomal abnormalities with diseases. Edvotek™ Chromosome Spread (Pre-Fixed Slides) helps students in staining and observe condensed chromosomes.
During mitosis, each of our chromosomes are duplicated. The chromosomes are then separated during mitosis, moving to opposite ends of the cell before cell division.
  • In this experiment cells have been arrested during metaphase and fixed to slides
  • Allows students to stain and observe the condensed chromosomes
  • Students will develop an understanding of karyotyping and the association of chromosomal abnormalities with diseases
  • For 6 Lab Groups
  • Complete in 35 minutes
  • Level: Beginner
Specifications
Material Glass
Mounting Type Whole Mount
Type Chromosome Spread (Pre-Fixed Slides)
Includes Instructions, pre-fixed chromosome spread slides, giemsa stain, mounting media, cover slips, transfer pipettes, immersion troughs.
Slide Type Prepared Slide