Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
pre-mRNA cleavage factor I (59 kDa subunit) Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10061600AF350
Description
pre-mRNA cleavage factor I (59 kDa subunit) Polyclonal antibody specifically detects pre-mRNA cleavage factor I (59 kDa subunit) in Human, Mammalia, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
pre-mRNA cleavage factor I (59 kDa subunit) | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CFIm59, cleavage and polyadenylation specific factor 7, 59kDa, CPSF 59 kDa subunit, FLJ12529,59 kDa subunit, FLJ39024, MGC9315, pre-mRNA cleavage factor I, 59 kDa subunit | |
The immunogen recognized by this antibody maps to a region between residue 175 and 225 of human cleavage and polyadenylation specificity factor 7, 59 kDa subunit using the numbering given in entry NP_079087.2 (GeneID 79869). | |
0.1 mL | |
Primary | |
Human, Mammalia, Primate | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
79869 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction