Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pre-Preserved Environmental Sample Containers

Catalog No. 12100261
Change view
Click to view available options
Color:
Amber
Clear
Preservative:
0.2 mL 1:1 Hydrochloric Acid
0.25 mL 1:1 Hydrochloric Acid
0.5 mL 1:1 Hydrochloric Acid
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Preservative
12-100-261 Amber 0.5 mL 1:1 Hydrochloric Acid
5005728130 Clear 0.5 mL 1:1 Hydrochloric Acid
12-100-255 Clear 0.2 mL 1:1 Hydrochloric Acid
12-100-183 Amber 0.2 mL 1:1 Hydrochloric Acid
12-100-251 Clear 0.25 mL 1:1 Hydrochloric Acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 12-100-261 Supplier Thermo Scientific™ Supplier No. PP14140AEP.5HA
Add to Cart
Add to Cart

Save the time, tedium and chemical contact associated with diluting and aliquoting chemical preservatives by using our chemically-preserved sample containers.

Simplify field preservation with Thermo Scientific™ Chemically-Preserved Environmental Sample Containers and enjoy peace of mind as well as convenience. Using certified containers and chemicals reduces the risk of container and preservative contamination. Let us save you the time, tedium and chemical contact associated with diluting and aliquoting chemical preservatives to prepare environmental sample container sets. Choose from our standard offering of chemically-preserved containers, or let us custom-create container/preservative combos according to your personal analytical needs.

  • Standard Thermo Scientific Certified sample containers chemically-preserved with certified chemical preservatives for environmental sample analyses
  • Packaged in compliance with the shipping requirements of 49 CFR Part 173.4 DOT Small Quantities Classification for highway and rail transport

Ordering Information:

Custom preserved containers available by special order:

  • Choose a standard certified Thermo Scientific environmental sample container
  • Choose a chemical from our standard chemical list
  • Specify the volume and concentration of chemical to aliquot into each container

Products available only in the US

Includes:

Certified sample containers dosed with chemical preservative; certificate of analysis for container and preservative, individually barcoded containers traceable to certs, sample analysis labels unattached for optional use, hazardous material compliant packaging. MSDS available upon request.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (English) Outer 1.1 in.
Color Amber
Diameter (Metric) Outer 28 mm
Material Glass (Type 1)
Volume (Metric) 40 mL
Height (English) 3.75 in.
Level Certified
No. per Case 72
Volume (English) 1 oz.
Volume (Metric) Total 40 mL
Closure Type Septa Cap
Thickness (Metric) 3.17 mm
Closure Size 24-414
Certifications/Compliance Certified to meet the EPA Performance Based Specifications for volatile organic compounds
Height (Metric) 95 mm
Preservative 0.5 mL 1:1 Hydrochloric Acid
Product Type Environmental Sample Container
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.