Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pre-Pro-Vasopressin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143640
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143640 Supplier Invitrogen™ Supplier No. PA5143640
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specific for locally produced levels of the ~ 20 kDa uncleaved pre-pro-vasopressin protein. Immunolabeling of the uncleaved pre-pro-vasopressin protein band is completely blocked by preadsorption of the antibody with the peptide used as antigen.

Vasopressin is a nine amino-acid peptide hormone that plays a key role in water and blood pressure homeostasis (Qureshi S., et al 2014). Vasopressin is the end-product of a highly processed 164 amino acid pre-pro-peptide. Processing of the vasopressin pre-pro-peptide results in three distinct peptides with a 1:1:1 ratio: vasopressin, neurophysin II, and copeptin (Arroyo J.P. et al, 2021). Vasopressin is the biologically active hormone, neurophysin II is a carrier molecule for vasopressin, and copeptin is the c-terminal glycosylated end-product. Vasopressin has been thought to be primarily made in the brain, and the sole source of vasopressin stimulating vasopressin V2 receptors in the kidney until recent studies (Arroyo JP, et al 2022). This antibody specially detects uncleaved pre-pro-vasopressin, accurately identifying locally produced vasopressin versus peripheral uptake of hypothalamic vasopressin or related peptides (Arroyo J.P. et al, 2022). This key research tool aided the Arroyo group in discovering immunoreactive vasopressin outside of the brain by recognizing localized vasopressin in human and mouse kidney, specifically in the distal nephron.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pre-Pro-Vasopressin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation 10mM HEPES with 100μg/mL BSA, 50% glycerol, 150mM NaCl and no preservative; pH 7.5
Gene AVP
Gene Accession No. P01185, P01186, P35455
Gene Alias ARVP; VP
Gene Symbols AVP
Host Species Rabbit
Immunogen Synthetic peptide corresponding to key amino acid residues surrounding the cleavage sites for the human vasopressin precursor peptide, conjugated to keyhole limpet hemocyanin (KLH).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11998, 24221, 551
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.