Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prealbumin PA Polyclonal Antibody, Huabio

Catalog No. 502544850
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4850 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4850 Supplier HUABIO RESEARCH INC Supplier No. 080910
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Prealbumin PA Polyclonal antibody specifically detects Prealbumin PA in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen Prealbumin PA
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TTR
Gene Accession No. P02766
Gene Alias AA408768; AI787086; Amyloidosis I; ATTR; carpal tunnel syndrome 1; CTS; CTS1; D17860; epididymis luminal protein 111; HEL111; HsT2651; Lr1; PALB; prealbumin; pre-albumin; prealbumin, amyloidosis type I; Tbpa; thyroxine-binding prealbumin; transthyretin; Tt; TTR
Host Species Rabbit
Immunogen Synthetic peptide within human Prealbumin PA aa 100-147.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7276
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.