Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PREB Antibody (SR1780), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322579100UL
Description
PREB Monoclonal antibody specifically detects PREB in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
PREB | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR1780 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
MGC3467, prolactin regulatory binding-element protein, prolactin regulatory element binding, prolactin regulatory element-binding protein, SEC12Mammalian guanine nucleotide exchange factor mSec12 | |
A synthesized peptide derived from human PREB (Uniprot number: Q9HCU5) | |
100 μL | |
Transcription Factors and Regulators | |
10113 | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction