Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PREB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553066
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553066 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553066 Supplier Invitrogen™ Supplier No. PA553066
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: REAHGIVVTD VAFLPEKGRG PELLGSHETA LFSVAVDSRC QLHLLPSRRS VP Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

Prolactin (PRL) expression in the pituitary is limited to specific cells. Pit-1 is a pituitary specific transcription factor that plays an important role in PRL expression, both in mature organism and during development. The PRL promoter contains numerous Pit-1 binding sites and these sites have been implicated in both basal level and kinase-mediated gene expression. The most proximal of these binding sites, termed 1P, has been shown to direct a response to numerous signals, such as cAMP and various G-proteins. A novel protein, termed PREB (prolactin regulatory element binding protein), has been recently identified that regulates PRL gene expression through the 1P site, though it contains no discernable DNA-binding motif. Recent studies suggest that PREB is encoded by a single-copy gene in both mice and humans and exhibits nuclear accumulation in pituitary cells. Evidence suggests that PREB is a novel transcription factor that assists in PRL expression whether alone, or in concert with Pit-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PREB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Preb
Gene Accession No. Q9HCU5, Q9WTV0, Q9WUQ2
Gene Alias C85705; mammalian guanine nucleotide exchange factor mSec12; PREB; prolactin regulatory binding-element protein; prolactin regulatory element binding; prolactin regulatory element binding protein; prolactin regulatory element-binding protein; Sec12
Gene Symbols Preb
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PREB. Recombinant protein control fragment (Product #RP-90969).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10113, 50907, 58842
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.