Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PREB Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30467320UL
Description
PREB Polyclonal specifically detects PREB in Human, Mouse samples. It is validated for Western Blot.Specifications
PREB | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Unconjugated | |
MGC3467, prolactin regulatory binding-element protein, prolactin regulatory element binding, prolactin regulatory element-binding protein, SEC12Mammalian guanine nucleotide exchange factor mSec12 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 13-202 of human PREB (NP_037520.1). | |
20 μL | |
Transcription Factors and Regulators | |
10113 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction