Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Precerebellin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520160
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520160 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520160 Supplier Invitrogen™ Supplier No. PA520160
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is mouse cerebellum tissue lysate. PA5-20160 can be used with blocking peptide PEP-0279.

Precerebellin is the precursor of the brain-specific hexadecapeptide cerebellin, a protein with substantial similarity to the globular region of the B chain of complement component C1q. Cerebellin exerts neuromodulatory functions by directly stimulating norepinephrine release via the adenylate cyclase/pka- dependent signaling pathway; and indirectly enhances adrenocortical secretion in vivo, through a paracrine mechanism involving medullary catecholamine release. The active form of precerebellin is highly enriched in postsynaptic structures of cerebellar Purkinje cells in cartwheel neurons of the dorsal cochlear nucleus. Because precerebellin belongs to the C1q/tumor necrosis factor superfamily of secreted proteins and has similarity to adiponectin and CTRP3, it has been suggested that precerebellin posseses functions other than those already stated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Precerebellin
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CBLN1
Gene Accession No. P23435, P63182, Q9R171
Gene Alias [des-Ser1]CER; [des-Ser1]-cerebellin; AI323299; Brain protein D3; Cbln1; CER; Cerebellin; cerebellin 1 precursor; cerebellin 1 precursor protein; cerebellin-1; des-Ser(1)-cerebellin; des-Ser1-cerebellin; Precerebellin
Gene Symbols CBLN1
Host Species Rabbit
Immunogen A 15 amino acid peptide from the middle of human precerebellin.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12404, 498922, 869
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.