Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Precipitated Sulfur, 99.5-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605785 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-785 12 kg Poly Pail
18-605-783 500 g Poly Bottle
18-605-784 2.5 kg Poly Pail
18-605-786 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-605-785 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S138412KG
Add to Cart
Add to Cart

S1384, 7704-34-9, S

Spectrum™ Chemical Precipitated Sulfur is an abundant, non-metal, multivalent chemical element. In cosmetics and personal care products Sulfur is used to enhance the appearance and feel of hair, by increasing hair body, suppleness, or sheen, or by improving the texture of hair that has been damaged physically or by chemical treatment and to enhance the appearance of dry or damaged skin by reducing flaking and restoring suppleness.

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
IUPAC Name sulfur
SMILES [S]

Specifications

CAS Max % 1
Quantity 12 kg
Percent Purity 99.5 to 100.5%
Grade Reagent
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.