Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B Medical Systems™ Precision Pharmacy Freezer, 337 L (11.9 ft.3)

Catalog No. BMSF380
Click to view available options
Capacity (Metric):
337 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Our Precision pharmacy freezers offer an economical option for reliable -32 to -20°C storage.

Premium and Precision Series laboratory and pharmacy freezers are medical-grade freezers designed to preserve temperature-sensitive enzymes, media, and clinical specimens.

  • Freezers include positive airflow rear wall plenums and interior designs, allowing ample clearance around inventory loads to assure uniform storage temperatures and overall stability
  • Advanced refrigeration systems include reserve refrigeration power for operation in high ambient installations to maintain temperature stability, uniformity, and fast recovery

Specifications

Type Lab Freezer
Capacity (English) 11.9 cu. ft.
Capacity (Metric) 337 L
Temperature Range -32°C to -20°C
Door Style Solid
No. of Doors 1 (1 compartment)
Insulation Polyurethane Foam
Interior Stainless Steel
Exterior Finish Painted Steel
No. of Drawers 9
Certifications/Compliance cUL, FDA Class I
Style Upright
Color Off-white
Condenser Type Air Cooled
Control Type Touch Screen
For Use With (Application) Lab and Pharmacy Storage
Warranty 5 Year Compressor, 2 Year Parts and Labor
Defrost Manual
Model F380
Dimensions (DxWxH) Exterior 25.6 x 23.7 x 76.8 in.
Depth (English) Exterior 25.6 in.
Height (English) Exterior 76.8 in.
Width (English) Exterior 23.7 in.
Monitoring Options B Connected
Access Security Alarm system
Backup Systems CO2 or LN2 Optional
Amperage 10 A
Plug Type NEMA 5-15
Voltage 115 V
Frequency 60 Hz
Phase 1
Wattage 360 w
Energy Usage 1.9 kWh/day
Compressor Bottom-mounted
Refrigerant R290
Shipping Weight (English) 368 lb.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.