Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Precision Plate Holder for Plates, for 7500 Fast System

Catalog No. 4359652
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-596-52 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-596-52 Supplier Applied Biosystems™ Supplier No. 4359652

This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or visit www.fishersci.com/quantstudio for more information and to contact a Life Science Specialist.

missing translation for 'validMainframeMsg'

Precision plate holder for 7500 fast system

Precision plate holder for 7500 fast system.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 7500 Fast Precision Plate Holder
Format 96-Well Plate
PCR Method qPCR
Reaction Speed Fast
For Use With (Equipment) 7500 Fast System
Product Type Precision Plate Holder
Quantity 1 each
Shipping Condition Room Temperature
For Use With (Application) Real Time PCR (qPCR)
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.