Learn More
Contec™ PREempt™ Concentrate Disinfectant
BIOPHARMA
Catalog No. p-6838121 Shop All Contec Inc. Products
Click to view available options
Volume (English):
1 gal.
55 gal.
Container Type:
Bottle
Drum
This item is not returnable. View return policy
This item is not returnable. View return policy
Works fast for large areas to ensure user, protocol and product compliance with fast contact times and broad-spectrum efficacy. Contec™ PREempt™ Concentrate Disinfectant requires dilution and is cost effective for large surfaces. PREempt Concentrate utilizes AHP™, a patented synergistic blend of commonly used, safe ingredients.
  • Cleaner - Effective disinfectant as well as a cleaner resulting in added confidence that disinfection can occur
  • Faster - Rapid and realistic contact times on a broad spectrum of difficult to kill pathogens
  • Responsible - Users are not exposed to VOCs (volatile organic compounds) and PPE is not required as per health and safety rating
  • Utilizes AHP™, a patented synergistic blend of commonly used, safe ingredients that when combined with low levels of hydrogen peroxide dramatically increase its potency and cleaning performance
  • 5 Minutes Bactericidal, Virucidal, Tuberculocidal, Fungicidal (1:16)
  • 3 minute Broad-Spectrum Sanitizing (1:128)
  • 30 second Broad-Spectrum Sanitizing (1:16)
Specifications
Product Type Concentrated Disinfectant Solution
Container Type Bottle
Volume (English) 1 gal.
Volume (Metric) 4 L
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.