Learn More
Contec™ PREempt™ RTU Disinfectant Solution
BIOPHARMA
Catalog No. 19039933 Shop All Contec Inc. Products
Click to view available options
Container Type:
Bottle
Pail
Spray bottle
Volume (English):
1 gal.
5 gal.
This item is not returnable. View return policy
This item is not returnable. View return policy
Cleans and disinfects environmental surfaces in cleanrooms and laboratory areas including work stations, fume hoods, equipment and other hard non-porous environmental surfaces. Contec™ PREempt™ RTU Disinfectant Solution helps to ensure user, protocol and product compliance with fast contact times and broad-spectrum efficacy.
  • AHP - Utilizes AHP™, a patented synergistic blend of commonly used, safe ingredients that when combined with low levels of hydrogen peroxide dramatically increase potency and cleaning performance
  • Cleaner - Effective disinfectant as well as a cleaner resulting in added confidence that disinfection can occur
  • Faster - Rapid and realistic contact times on a broad spectrum of difficult to kill pathogens
  • Responsible - Designed to be easier on surfaces, staff and occupants. Users are not exposed to VOCs (volatile organic compounds) and PPE is not required as per health and safety rating
  • 5 minute tuberculocidal
  • 10 minute fungicidal
  • 30 seconds broad-spectrum sanitizing
  • 1 minute bactericidal, virucidal
Specifications
Product Type Disinfectant Solution
Container Type Spray bottle
Volume (Metric) 0.9 L
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.