Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pref-1/DLK1/FA1 Antibody (1168B), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB8634MFV450
Description
Pref-1/DLK1/FA1 Monoclonal antibody specifically detects Pref-1/DLK1/FA1 in Mouse samples. It is validated for Western Blot, Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
| Pref-1/DLK1/FA1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Signal Transduction | |
| 8788 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| 1168B | |
| Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry | |
| delta-like 1 homolog (Drosophila), DLK, DLK-1, FA1fetal antigen 1, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG | |
| Mouse myeloma cell line NS0-derived recombinant mouse Pref-1/DLK1/FA1, Ala24-Gln305, Accession # Q09163 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction