Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pref-1/DLK1/FA1 Antibody (MM0514-9D8), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211855AF405
Description
Pref-1/DLK1/FA1 Monoclonal specifically detects Pref-1/DLK1/FA1 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
Pref-1/DLK1/FA1 | |
Monoclonal | |
Alexa Fluor 405 | |
P80370 | |
DLK1 | |
Human recombinant Pref-1/DLK1/FA1 | |
0.1 mL | |
Signal Transduction | |
8788 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Flow Cytometry | |
MM0514-9D8 | |
Western Blot, Flow Cytometry | |
delta-like 1 homolog (Drosophila), DLK, DLK-1, FA1fetal antigen 1, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction