Learn More
Invitrogen™ Plasma Kallikrein Monoclonal Antibody (13G11)

Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Plasma Kallikrein |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 13G11 |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Klkb1 |
| Gene Accession No. | P03952 |
| Gene Alias | antigen, prostate specific; APS; EC 3.4.21.34; Fletcher factor; Kal3; Kal-3; kallikrein; kallikrein 3, plasma; kallikrein B plasma; kallikrein B, plasma (Fletcher factor) 1; kallikrein B, plasma 1; kallikrein B1; Kalp1; KALP15; kininogenin; Klk3; KLKB1; Pk; PKK; PKKD; plasma kallikrein; Plasma kallikrein heavy chain; Plasma kallikrein light chain; Plasma prekallikrein; PPK; prekallikrein; PSA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.