Learn More
Invitrogen™ PRELID1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531087
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (99%), Rat (99%), Pig (97%), Chicken (88%), Rhesus Monkey (99%), Chimpanzee (98%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be important for the development of vital and immunocompetent organs.
Specifications
PRELID1 | |
Polyclonal | |
Unconjugated | |
PRELID1 | |
25 kDa protein of relevant evolutionary and lymphoid interest; 2610524G07Rik; CGI-106; Preli; PRELI domain containing 1; PRELI domain-containing protein 1, mitochondrial; PRELID1; protein of relevant evolutionary and lymphoid interest; PX19; PX19 homolog; px19-like protein; RGD1308082; SBBI12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27166 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y255 | |
PRELID1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 189 of Human PRELID1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.