Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRELID1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552187
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552187 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552187 Supplier Invitrogen™ Supplier No. PA552187
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VAHSVYVLED SIVDPQNQTM TTFTWNINHA RLMVVEERCV YCVNSDNSGW TEIRREAWVS SSLFGVSRAV QEFGLARFKS NVTKTMKGFE YILAKLQGEA PSKTLVETAK EAKEKAKETA LAATEKA Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

May be important for the development of vital and immunocompetent organs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRELID1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRELID1
Gene Accession No. Q9Y255
Gene Alias 25 kDa protein of relevant evolutionary and lymphoid interest; 2610524G07Rik; CGI-106; Preli; PRELI domain containing 1; PRELI domain-containing protein 1, mitochondrial; PRELID1; protein of relevant evolutionary and lymphoid interest; PX19; PX19 homolog; px19-like protein; RGD1308082; SBBI12
Gene Symbols PRELID1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRELID1. Recombinant protein control fragment (Product #RP-102152).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27166
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.