Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRELID1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 108771AP150UL

Description
May be important for the development of vital and immunocompetent organs.Specifications
PRELID1 | |
Polyclonal | |
Unconjugated | |
PRELID1 | |
CGI 106, PRELI, PRELI domain containing 1, PRELID1, PX19, Px19 like protein, SBBI12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27166, 290995, 66494 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8R107, Q9Y255 | |
PRELID1 | |
PRELID1 Fusion Protein Ag1037 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction