Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRELID2 Monoclonal Antibody (OTI3B7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA810710

Description
PRELID2 (PRELI Domain Containing 2) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include phosphatidic acid transporter activity. An important paralog of this gene is PRELID3B.Specifications
PRELID2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8N945 | |
PRELID2 | |
Full length human recombit protein of human PRELID2 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI3B7 | |
Unconjugated | |
PRELID2 | |
PRELI domain containing 2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
153768 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction