Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRELP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120649
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120649 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120649 Supplier Invitrogen™ Supplier No. PA5120649
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse lung, Rat heart. Immunogen sequence: LDLQHNRLSD GVFKPDTFHG LKNLMQLNLA HNILRKMPPR VPTAIHQLYL DSNKIETIPN GYFKSFPNLA FIRLNYNKLT DRGLPKNSFN ISNLLVLHLS HNRISSVPAI NNRLEHLYLN NNSIEKINGT QICPNDLVAF HDFSSDLENV PHLRYLRLDG N.

PRELP is a leucine-rich repeat protein present in connective tissue extracellular matrix. This protein functions as a molecule anchoring basement membranes to the underlying connective tissue. This protein has been shown to bind type I collagen to basement membranes and type II collagen to cartilage. It also binds the basement membrane heparan sulfate proteoglycan perlecan. This protein is suggested to be involved in the pathogenesis of Hutchinson-Gilford progeria (HGP), which is reported to lack the binding of collagen in basement membranes and cartilage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRELP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PRELP
Gene Accession No. Q9EQP5, Q9JK53
Gene Alias 55 kDa leucine-rich repeat protein of articular cartilage; 7330409J17Rik; MST161; MSTP161; PRELP; Prolargin; prolargin proteoglycan; proline and arginine rich end leucine rich repeat protein; proline arginine-rich end leucine-rich repeat; proline/arginine-rich end leucine-rich repeat protein; proline-arginine-rich end leucine-rich repeat protein; SLRR2A
Gene Symbols PRELP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-360 of human PRELP (NP_002716.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116847, 84400
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.