Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meridian Bioscience Premier HpSA Flex

Click to view available options
Detectable Analytes:
H. pylori
Description
- Premier™ HpSA™ Flex is an enzyme immunoassay with high-volume throughput approved for fresh (unpreserved) and preserved stool specimens
- Aligns with the AGA and ACG guidelines for H. pylori non-invasive active infection testing
- ELISA technology lends itself to highvolume throughput
- Detachable microwells maximize determinations per kit utilization
- Can be read visually or on automated systems
- Approved for fresh unpreserved stool and preserved stool specimens in Cary-Blair or C and S
- Flexible options for stool transport vials can improve laboratory efficiencies, preserve sample integrity, and streamline sample processing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
CE Marker | No |
Clia Complexity | High |
Detectable Analytes | H. pylori |
Sample Type | Preserved (Cary-Blair or C and S) Stool or Fresh Stool (Unpreserved) |
Type | Immunoassay |
Disposable | Single-use |
Form | Ready-to-Use (RTU) |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction