Learn More
Four E's Healthcare Premium SMS Lab Coat with Anti-Static Coating
Catalog No. p-7220868 Shop All Fisherbrand Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Premium knee-length lab coat from Four E's Healthcare made of 3-layer breathable SMS fabric with anti-static coating, offers superior comfort for extended use and protects from low fluid levels
  • Anti-static SMS lab coats with knit collars, cuffs and snap front with five buttons
  • Protective 3-ply breathable SMS fabric for comfort and protection
  • Offers protection from low fluid levels
    • SMS fabric material meets AAMI Level 2 liquid barrier performance
  • Color: blue
  • Three reinforced pockets (two hip pockets, one breast pocket)
  • Comes with anti-static coating
    • Keeps wearer safe in static-prone environments and avoids clinging
Specifications
Product Type General Purpose Lab Coat
Size Small
Gender Unisex
Material SMS
Color Blue
Closure Type Snaps
Cuff Style Knit
Sterility Non-sterile
Resistance Fluids
No. of Pockets 3
Collar Type Knit
Latex Free Yes
Model 4ESMSLC45
Certifications/Compliance AAMI Level 2
Construction 3-ply SMS fabric with anti-static coating
Show More Show Less
Please note: The SMS fabric is fluid resistant, and meets AAMI Level 2 Liquid barrier performance. However, due to the snap front design, the coat does not qualify for AAMI PB70 standard testing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.