Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PREP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547611
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547611 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547611 Supplier Invitrogen™ Supplier No. PA547611
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse PREP is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

p33cdk2 associates with cyclin A in human cells. Kinase activity associated with cyclin A-cdc2 is found only in G2-phase. Cdk2 also complexes with cyclins E, D1, and D3. Cyclin E-cdk2 kinase is active in the G1- and S-phases of the cell cycle and is important (as does cyclin A-cdk2) for the progression from G1-to S-phase. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PREP
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene PREP
Gene Accession No. P48147
Gene Alias AI047692; AI450383; D10Wsu136e; dJ355L5.1 (prolyl endopeptidase); EC 3.4.21.26; MGC16060; PE; PE {ECO:0000250; PEP; Pop; post-proline cleaving enzyme; post-proline cleaving enzyme {ECO:0000250; Prep; prep {ECO:0000312; Prolyl endopeptidase; prolyl endopeptidase {ECO:0000250; prolyl oligopeptidase; RGD:620841}; rPop; UniProtKB:P23687}
Gene Symbols PREP
Host Species Goat
Immunogen Sf 21-derived recombinant human PREP Leu2-Pro710.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5550
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.