Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PREP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598086
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598086 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598086 Supplier Invitrogen™ Supplier No. PA598086
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PREP Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

p33cdk2 associates with cyclin A in human cells. Kinase activity associated with cyclin A-cdc2 is found only in G2-phase. Cdk2 also complexes with cyclins E, D1, and D3. Cyclin E-cdk2 kinase is active in the G1- and S-phases of the cell cycle and is important (as does cyclin A-cdk2) for the progression from G1-to S-phase. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PREP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PREP
Gene Accession No. P48147
Gene Alias AI047692; AI450383; D10Wsu136e; dJ355L5.1 (prolyl endopeptidase); EC 3.4.21.26; MGC16060; PE; PE {ECO:0000250; PEP; Pop; post-proline cleaving enzyme; post-proline cleaving enzyme {ECO:0000250; Prep; prep {ECO:0000312; Prolyl endopeptidase; prolyl endopeptidase {ECO:0000250; prolyl oligopeptidase; RGD:620841}; rPop; UniProtKB:P23687}
Gene Symbols PREP
Host Species Rabbit
Immunogen Recombinant Human Prolyl endopeptidase protein (539-691AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5550
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.