Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fischer Technical Company™ Prepared Culture Media in Plates
Shop All Fischer Technical Company Products

Click to view available options
Media Type:
LB Agar
MacConkey Agar
Nutrient Agar
Plate Count Agar
Potato Dextrose Agar
Sabouraud Dextrose Agar
Starch Agar
Tryptic Soy Agar
Detects:
Bacteria
Exoenzymes
Fungi
Gram Negative Bacteria
Microbes
Non-fastidious Aerobic and Anaerobic Microorganisms
Non-fastidious Microorganisms
Yeast, Mold
Description
- Sterile plates are ready for immediate use
- Supplied in disposable plastic petri dishes
- Packed in clear, heat-sealed cellophane bags
Specifications
Specifications
Detects | Exoenzymes |
Format | Plate |
Manufacturing Origin | US with Globally Sourced Materials |
Media Type | Starch Agar |
Product Type | Agar Plate |
Sterility | Sterile |
Target Organism Class | Bacteria |
Test Type | Microbiological Tests |
For Use With (Application) | Microbiological Testing |
Quantity | 10 Plates |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction