Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Capsella Embryo, Longitudinal Section
SCI_ED

Catalog No. S30382 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30382 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30382 Supplier Eisco™ Supplier No. BS18241

Prepared microscope slide of a longitudinal section of mature embryo of Sheperd's Purse (Capsella). Stained for better visualization of bending cotyledons inside the mature embryo. Great choice for biology classrooms.

  • Pre-cleaned glass slide with ground edges
  • Size: 1 x 3 in. (25 x 77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSS standards
  • Showing bending cotyledons inside the mature embryo

Specifications

Description Longitudinal section of a Sheperd's Purse -Capsella mature embryo, with visible bending cotyledons inside
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1
Specimen Type Capsella Embryo-Mature
Size 1 x 3 in.
Slide Type Prepared Slide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.