Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PREPL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564098
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564098 Supplier Invitrogen™ Supplier No. PA564098
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PQHYPSIHIT AYENDERVPL KGIVSYTEKL KEAIAEHAKD TGEGYQTPNI ILDIQPGGNH VIEDSHKKIT AQIKFLYE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.

The protein encoded by this gene belongs to the prolyl oligopeptidase subfamily of serine peptidases. Mutations in this gene have been associated with hypotonia-cystinuria syndrome, also known as the 2p21 deletion syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PREPL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Prepl
Gene Accession No. Q4J6C6
Gene Alias 2810457N15Rik; 9530014L06Rik; D030028O16Rik; KIAA0436; mKIAA0436; Prepl; prolyl endopeptidase; prolyl endopeptidase-like; Prolylendopeptidase-like; putative prolyl oligopeptidase; RGD1310143
Gene Symbols Prepl
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PREPL. Recombinant protein control fragment (Product #RP-95839).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9581
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.