Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PREPL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589772
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589772 Supplier Invitrogen™ Supplier No. PA589772
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AVTLEAPFLD VLNTMMDTTL PLTLEELEEW GNPSSDEKHK NYIKRYCPYQ NIKPQHYPSI HITAYENDER VPLKGIVSYT EKLKEAIAEH AKDTGEGYQT PNIILDIQPG GNHVIEDSHK KITAQIKFLY EELGLDSTSV FEDLKKYLKF; Positive Samples: U-87MG, Mouse brain, Rat brain; Cellular Location: Cytoplasm, cytosol.

The protein encoded by this gene belongs to the prolyl oligopeptidase subfamily of serine peptidases. Mutations in this gene have been associated with hypotonia-cystinuria syndrome, also known as the 2p21 deletion syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PREPL
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Prepl
Gene Accession No. Q4J6C6, Q5HZA6, Q8C167
Gene Alias 2810457N15Rik; 9530014L06Rik; D030028O16Rik; KIAA0436; mKIAA0436; Prepl; prolyl endopeptidase; prolyl endopeptidase-like; Prolylendopeptidase-like; putative prolyl oligopeptidase; RGD1310143
Gene Symbols Prepl
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 578-727 of human PREPL (NP_0060272).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213760, 298771, 9581
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.