Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PREPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 124781AP150UL

Description
The protein encoded by this gene belongs to the prolyl oligopeptidase subfamily of serine peptidases. Mutations in this gene have been associated with hypotonia-cystinuria syndrome, also known as the 2p21 deletion syndrome.Specifications
PREPL | |
Polyclonal | |
Unconjugated | |
PREPL | |
KIAA0436, PPCEL, PREPL, prolyl endopeptidase like, Prolylendopeptidase like | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
213760, 298771, 9581 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4J6C6, Q5HZA6, Q8C167 | |
PREPL | |
PREPL Fusion Protein Ag3196 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction