Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Presenilin-2 Antibody (APS 21), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10074512
Description
Presenilin-2 Monoclonal specifically detects Presenilin-2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Presenilin-2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100 - 1:1000, ELISA 4.5 ug/ml, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 0.75 ug/ml, Immunohistochemistry-Paraffin 1:100 | |
AD3LP, AD4AD3L, AD5, E5-1, EC 3.4.23, EC 3.4.23.-, presenilin 2 (Alzheimer disease 4), presenilin-2, PS-2, PS2CMD1V, PSNL2, STM2STM-2 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Presenilin 2 (APS 21) | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
APS 21 | |
Unconjugated | |
PBS with 1mg/ml BSA with 0.05% Sodium Azide | |
PSEN2 | |
Synthetic peptide corresponding to residues C(31) Q E G R Q G P E D G E N T A(45) of human PS2. | |
200 μg | |
Alzheimers Research, Apoptosis, Neuroscience | |
5664 | |
Human, Mouse, Rat, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction