Learn More
Presenilin-2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | Presenilin-2 |
---|---|
Applications | Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-7475
![]() |
Bethyl Laboratories, Inc
A304342AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
Familial Alzheimer's disease is often characterized by an early (<60 years of age) and rapid deterioration of the central nervous system. The symptoms are caused by the abnormal buildup of senile plaques composed of the 42 residue amyloid-beta peptide. This peptide is the result of the amyloid precursor protein being cleaved by the presenilin-gamma-secretase complex. Recent studies have linked mutations in presenilin 1 (PS1) and presenilin 2 (PS2) to a rapid increase in plaque accumulation. PS1 and PS2 are integral membrane proteins that contain 6-8 transmembrane domains and are predomitly localized within the endoplasmic reticulum and Golgi of neurons within the brain. Within the transmembrane regions of PS1 and PS2 there is over 60% homology, with the largest divergence found at the N-terminus and large loop region.Specifications
Presenilin-2 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
PS2, AD4, Alzheimer disease 4, PS2, PSEN 2, STM2 | |
PSEN2 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P49810 | |
5664 | |
Between 275 and 325 | |
Primary | |
4° C | |
PSEN2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.