Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Presenilin-2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501567473
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7473 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7473 Supplier Bethyl Laboratories, Inc Supplier No. A304342A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine, Pig and Orangutan

Familial Alzheimer′s disease is often characterized by an early (<60 years of age) and rapid deterioration of the central nervous system. The symptoms are caused by the abnormal buildup of senile plaques composed of the 42 residue amyloid-beta peptide. This peptide is the result of the amyloid precursor protein being cleaved by the presenilin-gamma-secretase complex. Recent studies have linked mutations in presenilin 1 (PS1) and presenilin 2 (PS2) to a rapid increase in plaque accumulation. PS1 and PS2 are integral membrane proteins that contain 6-8 transmembrane domains and are predomitly localized within the endoplasmic reticulum and Golgi of neurons within the brain. Within the transmembrane regions of PS1 and PS2 there is over 60% homology, with the largest divergence found at the N-terminus and large loop region.

Specifications

Antigen Presenilin-2
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene PSEN2
Gene Accession No. P49810
Gene Alias PS2, AD4, Alzheimer disease 4, PS2, PSEN 2, STM2
Gene Symbols PSEN2
Host Species Rabbit
Immunogen Between 275 and 325
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5664
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.