Learn More
Restek™ Presents SGE Replacement Needles for Varian 8000 Series Autosampler Syringes
SDP
Catalog No. 06712519 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Needle Point Style:
Side Hole
Side Hole, Tapered Needle Shaft
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Point Style
06-712-519 Side Hole
06-712-518 Side Hole, Tapered Needle Shaft
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-712-519 Supplier Restek™ Supplier No. 24857
For Bruker/Varian 8000 Series Autosampler Syringes.
  • Volume: 10μL
  • Needle Length: 53mm
Specifications
Needle Gauge 23 G
Length (English) Needle 2.1 in.
Product Type Replacement Needle
Needle Point Style Side Hole
For Use With (Equipment) Varian 8000 Series Autosampler