Learn More
Carolina™ Formalin Preserved Sheep Half Brain
Shop All Carolina Biological Supply Products
Click to view available options
Quantity:
1/Pk.
Description
- Similar to human brain
- Sheep brain is shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
- Odorless fluid designed to minimize unpleasant odor of formaldehyde
- Plain: No color injection
- Individually damp-packed in vacuum-sealed bags
Specifications
Specifications
| Type | Preserved Specimen |
| Preservative | Formalin |
| Injection Type | Plain |
| Quantity | 1/Pk. |
| Packaging | Bag |
| Autoclavable | Not Autoclavable |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov