Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PrestoBlue™ and PrestoBlue™ HS Cell Viability Reagents

Catalog No. p-200102300
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
Purity or Quality Grade:
HS Purified
Standard
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Purity or Quality Grade
A13261 25 mL Standard
A13262 100 mL Standard
P50200 25 mL HS Purified
P50201 100 mL HS Purified
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A13261 Supplier Invitrogen™ Supplier No. A13261
Only null left
Add to Cart
Edge
Add to Cart

Monitor cell proliferation and viability with the resazurin-based PrestoBlue and PrestoBlue HS cell viability reagents. These nontoxic and cell permeable reagents quantitatively measure cell viability in as little as 10 minutes using absorbance- or fluorescence-based microplate readers.

PrestoBlue and PrestoBlue HS cell viability reagents are ready-to-use, nontoxic, resazurin-based solutions that function as long term kinetic cell health indicators for fluorescence and absorbance-based microplate assays. These reagents are cell permeable and use the reducing power of living cells to quantitatively measure cell proliferation and viability in as little as 10 minutes. PrestoBlue HS Cell Viability Reagent is an improved version of PrestoBlue reagent, containing highly purified resazurin, which increases sensitivity and reduces background in cell proliferation assays. Add the power of multiplexing with reagents such as the CyQUANT Direct assay to improve your data analysis.

Measuring changes in cell viability is a fundamental method for assessing cell health, determining genotoxicity, and evaluating anti-cancer drugs. Cell health can be monitored by detecting changes in several key indicators, including changes to plasma membrane integrity, DNA synthesis, DNA content, enzyme activity, presence of ATP, and cellular reducing environment.

Features of PrestoBlue and PrestoBlue HS cell viability reagents include:

  • Optimized for reliable detection of mammalian cell viability in only 10 minutes
  • Measure cell viability and proliferation in various cell types, including mammalian (2D and 3D), bacteria, and yeast cells
  • A convenient add-and-read format—no mixing, washing, or cell lysis required
  • Compatible with either fluorescence- or absorbance-based microplate readers
  • Obtain high quality cell proliferation results with a large dynamic range
  • Monitor living cells using real time kinetics and continue culturing them for downstream multiplexed functional assays

Quick 10-minute incubations

Detect mammalian cell viability after just a 10-minute incubation with PrestoBlue and PrestoBlue HS cell viability reagents. These are the only live-cell viability reagents on the market that provide robust data in 10 minutes following reagent addition. All other resazurin-based cell viability reagents require a 1–4 hour incubation with the reagent prior to obtaining data. For best results using 3D samples, it is recommended to incubate the samples for 18–24 hours prior to measurements.

Exceptional sensitivity and accuracy

PrestoBlue and PrestoBlue HS cell viability reagents can detect as few as 12 cells per well in a 384-well plate, depending on the cell type and incubation time. After a 10-minute incubation, PrestoBlue reagent can linearly detect cells down to 98 cells/well with a Z' value greater than 0.5.

Simplified assay format

PrestoBlue and PrestoBlue HS cell viability reagents are addition-only reagents, eliminating the need for cell lysis or solubilization steps required by other assays. This minimizes hands-on time and simplifies the process.

Nontoxic live cell assays

PrestoBlue and PrestoBlue HS cell viability reagents are nontoxic and do not require cell lysis, allowing cells to be incubated for up to 24 hours. Since no lysis is required, the diluted PrestoBlue or PrestoBlue HS solution can be removed from the cells and replaced with complete growth medium for further culturing and use in downstream assays.

PrestoBlue HS reagent—a highly purified version of PrestoBlue reagent with improved performance

All resazurin-based reagents contain a detectable amount of resorufin contamination due to the synthesis and manufacturing process, which contributes to high background fluorescence and reduces performance in cell viability assays. To reduce background and improve performance, an innovative purification process was developed to remove the contaminating resorufin, resulting in the highly pure resazurin used in the PrestoBlue HS Cell Viability Reagent. This highly purified, nontoxic resazurin reduces background fluorescence by >50% and increases signal-to-background by >100% in cell viability microplate assays.

Features of PrestoBlue HS Cell Viability Reagent include:

  • Removal of contaminants from resazurin, resulting in >50% reduction in background fluorescence
  • Increased signal-to-background ratio by >100%, providing a large assay signal window
  • Offers high sensitivity with a linear response, detecting as few as 10 cells per well

Mechanism of PrestoBlue and PrestoBlue HS reagents

Using resazurin-based reagents to monitor changes in cellular reducing environment or metabolic activity is a well-established and reliable method to indicate cell viability or death. PrestoBlue and PrestoBlue HS reagents contain resazurin, which is a nontoxic, cell permeable compound that is blue in color and virtually nonfluorescent. When added to cells, the resazurin-based PrestoBlue and PrestoBlue HS reagents enter cells and are modified by the reducing environment of viable cells. In live cells, resazurin is reduced to resorufin, a compound that is red in color and highly fluorescent. Viable cells continuously convert resazurin to resorufin, increasing the overall fluorescence of the medium surrounding the cells. The conversion of resazurin to resorufin also results in a pronounced color change, allowing cell viability to be detected using either absorbance- or fluorescence-based plate readers.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Bacteria, Eukaryotic Cells, Fungal Cells
Color Blue
Concentration 10X
Content And Storage Store at 2°C to 8°C and protect from light.
Detection Method Colorimetric, Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Microplate Reader, Spectrophotometer, HTS Reader
Product Type Cell Viability Reagent
Dye Type Resazurin
Emission 590 to 615
Excitation Wavelength Range 535-560 nm
Form Liquid
Format 96-well plate, Cuvettes, 384-well plate
Incubation Time 10 min. to 2 hr.
Product Line PrestoBlue
Purity or Quality Grade Standard
Quantity 25 mL
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Throughput High-throughput Compatible
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.