Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Presumptive Blood Test Kit
GREENER_CHOICESCI_ED

Catalog No. s96638 Shop All Aldon Products
Change view
Click to view available options
Class Size:
30 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S96638 30 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Phenolphthalein and Methanol, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S96638 Supplier Innovating Science™ Supplier No. IS9002
Only null left
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Phenolphthalein and Methanol, which are known to the State of California to cause cancer and reproductive harm.

Includes

Instructions, phenolpthalein (2% solution UN1814, 2 x 25 mL), ethyl alcohol UN1170 (2 x 25 mL), hydrogen peroxide (3% solution, 50 mL), five blood standard strips, 50 cotton swabs. Enough materials for 30 tests.

Use phenolphthalein to test for the presence of blood.

Use swabs to collect samples of suspected blood evidence for testing.

Does not distinguish between animal and human blood; further serology tests would be needed.

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13+ years
For Use With (Application) For presumptive blood test
Includes 50 mL phenolpthalein, 2% solution un1814, 50 mL ethyl alcohol un1170, 50 mL hydrogen peroxide, 3% solution, 5 blood standard strips, 50 cotton swabs, teacher manual and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 4.25 x 4.625 x 5.5 in.
Class Size 30 Students
Product Type Presumptive Blood Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.