Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pretend and Play™ Healthy Food Sets
SCI_ED

Catalog No. p-3492671
Click to view available options
Product Type:
Pretend and Play™ Healthy Breakfast Food Set
Pretend and Play™ Healthy Dinner Set
Pretend and Play™ Healthy Lunch Set
Includes:
10.75 x 5.25 x 8.5 in. shopping basket and an activity guide, english muffin half, egg, banana, 7 strawberries in a bowl, 2 pancakes, yogurt cup and orange juice
10.75 x 5.25 x 8.5 in. shopping basket, 2 slices of wheat bread, 2 slices of turkey, lettuce, tomato, cheese, 4 celery sticks, 2 pretzels, grapes, orange and milk, activity card and teaching guide
10.75 x 5.25 x 8.5 in. shopping basket, grilled chicken breast, spaghetti with tomato sauce, peas, beans, potato, 4 apple slices, 5 carrot sticks, french bread, milk, handy basket and teaching guide
CHOKING HAZARD —Small parts. Not for children under 3 yrs.
CHOKING HAZARD —Small parts. Not for children under 3 yrs.

Includes

  • Shopping Basket: 10.8 x 5.25 x 8.5 in. (27.3 x 13.3 x 21.6cm)
  • Activity card
  • Teaching guide

Encourage healthy choices with this realistic-looking set of nutritious foods,

  • Great for developing oral language through role play in early childhood and ELL classrooms
  • All pieces are actual size and washable

  • Breakfast set includes an English muffin half, egg, banana, seven strawberries in a bowl, two pancakes, butter, yogurt cup, and orange juice
  • Lunch set includes two slices of wheat bread, two slices of turkey, lettuce, tomato, cheese, four celery sticks, two pretzels, grapes, orange, and milk
  • Dinner set includes grilled chicken breast, spaghetti with tomato sauce, peas, beans, potato, four apple slices, five carrot sticks, French bread, and milk

Order Info

Grades PK-K

Specifications

Product Type Pretend and Play™ Healthy Breakfast Food Set
Grade Pre-K to K
Age 3
For Use With (Application) Help students learn to make good nutritional choices by starting each day with a healthy breakfast.
Includes 10.75 x 5.25 x 8.5 in. shopping basket and an activity guide, english muffin half, egg, banana, 7 strawberries in a bowl, 2 pancakes, yogurt cup and orange juice
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.